Learn More
Medchemexpress LLC Tenacissoside G | 191729-43-8 | MFCD22417326 | 99.6% | 792.95 | C42H64O14 | 10 MG
SDP
Catalog No. 5000296135
Encompass_Preferred
Tenacissoside G is a C21 steroid isolated from the stems of Marsdenia tenacissima. It is reported to reverse multidrug resistance in P-glycoprotein (Pgp/ABCB1/MDR1)-overexpressing cancer cells and is used as a research reagent for studying P-gp-mediated drug transport and resistance mechanisms.
  • natural C21 steroid isolated from plant stems.
  • reverses P-glycoprotein-mediated multidrug resistance in cell models.
  • used in research on drug transport, resistance pathways, and biochemical assays.
  • provided in small, defined mass amounts suitable for in vitro studies.
  • characterized by CAS 191729-43-8 and molecular weight 792.95.
Catalog No. 50-002-96135 Supplier Medchemexpress LLC Supplier No. HYN210310MG
Add to Cart
Add to Cart
Tenacissoside G is a C21 steroid isolated from the stems of Marsdenia tenacissima. It is reported to reverse multidrug resistance in P-glycoprotein (Pgp/ABCB1/MDR1)-overexpressing cancer cells and is used as a research reagent for studying P-gp-mediated drug transport and resistance mechanisms.
  • natural C21 steroid isolated from plant stems.
  • reverses P-glycoprotein-mediated multidrug resistance in cell models.
  • used in research on drug transport, resistance pathways, and biochemical assays.
  • provided in small, defined mass amounts suitable for in vitro studies.
  • characterized by CAS 191729-43-8 and molecular weight 792.95.