Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tenacissoside i | 191729-44-9 | 5 MG
SDP

Catalog No. 5000421696
Encompass_Preferred

Tenacissoside I is a C21 steroid sourced from the Marsdenia tenacissima plant, where it is found at relatively high levels. This product is intended for research use only.

  • Purity: 99.0%
  • Molecular formula: C44H62O14
  • Molecular weight: 814.95
  • Appearance: Solid, white to off-white
  • Structure classification: Steroids
  • Initial source: Plants other families
  • Storage conditions: 4°C, sealed storage, away from moisture and light. In solvent: -80°C for 6 months; -20°C for 1 month.

Catalog No. 50-004-21696 Supplier Medchemexpress LLC Supplier No. HYN21025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Tenacissoside I is a C21 steroid sourced from the Marsdenia tenacissima plant, where it is found at relatively high levels. This product is intended for research use only.

  • Purity: 99.0%
  • Molecular formula: C44H62O14
  • Molecular weight: 814.95
  • Appearance: Solid, white to off-white
  • Structure classification: Steroids
  • Initial source: Plants other families
  • Storage conditions: 4°C, sealed storage, away from moisture and light. In solvent: -80°C for 6 months; -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.