Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tenacity No. 125 Flux Powder, working range 750-1200°C (1382-2102°F)

Catalog No. AA4729934
Change view
Click to view available options
400 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA4729934 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4729934 Supplier Thermo Scientific™ Supplier No. 4729934

Tenacity No.125 Flux Powder is suitable for use on copper and copper based alloys, mild and low alloy steels and tungsten carbide.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Tenacity No.125 Flux Powder is suitable for use on copper and copper based alloys, mild and low alloy steels and tungsten carbide.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 400 g
Form Solid
UN Number UN3288
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.