Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ TenActiv™ Knit Cut-Resistant Gloves, Polyurethane Coating

Catalog No. 17400211
Change view
Click to view available options
Size:
5
6
7
8
9
10
11
12
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-400-211 12
17-400-204 5
17-400-205 6
17-400-206 7
17-400-207 8
17-400-208 9
17-400-209 10
17-400-210 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-400-211 Supplier Superior Glove™ Supplier No. STAGHVPU12
Add to Cart
Add to Cart

Knit, cut-resistant gloves with ANSI Cut Level A5 and polyurethane coating on palms and fingers.

A hi-vis knit shell gives these gloves an outstanding level of visibility on job sites, and their stronger-than-steel TenActiv™ yarn ensures ANSI Level A5 cut protection. Their shell has been made lint-free to avoid contaminants, and a polyurethane coating at the palms provides fingers with a tactile feel.

  • Made with high-strength TenActiv™ yarn that is stronger than steel on an equal weight basis
  • Polyurethane coating offers excellent dexterity and strong grip

Specifications

Coating Material Polyurethane
Color Hi-vis Yellow
Cut Resistance Rating ANSI A5
Material TenActiv Yarn
Size 12
For Use With (Application) Glass Manufacturing, Metal Fabrication, Automotive Industry
Puncture Rating ANSI/ISEA 105-16 Level 4
Yarn Gauge 13
Length (English) Cuff 2 in.
Length (Metric) Cuff 5.1 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.