Learn More
TARGETMOL CHEMICALS INC TENAPANOR 10MG
SDP
Catalog No. 502576305
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tenapanor (RDX 5791) is an inhibitor of the sodium-proton (Na(+)/H(+)) exchanger NHE3 which plays a prominent role in sodium handling in the gastrointestinal tract and kidney. Tenapanor possesses an excellent preclinical safety profile and as of now there are no serious concerns about its side effects. purity: 99%
Catalog No. 50-257-6305 Supplier TARGETMOL CHEMICALS INC Supplier No. T758710MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tenapanor (RDX 5791) is an inhibitor of the sodium-proton (Na(+)/H(+)) exchanger NHE3 which plays a prominent role in sodium handling in the gastrointestinal tract and kidney. Tenapanor possesses an excellent preclinical safety profile and as of now there are no serious concerns about its side effects. purity: 99%