Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tenapanor hydrochloride | 1234365-97-9 | MFCD31631378 | 99.6% | 1217.97 | C50H68Cl6N8O10S2 | 10 MG
SDP

Catalog No. 5000285550
Encompass_Preferred

Tenapanor hydrochloride is a potent, orally active sodium/hydrogen exchanger 3 (NHE3) inhibitor supplied for research use. It is provided as a white to off-white solid and is available in solid and solution formats for in vitro and in vivo studies.

  • Potent NHE3 inhibitor for pharmacology and preclinical research.
  • Provided as a white to off-white solid and as solution formulations.
  • Reported purity 99.6% with analytical data (COA, HPLC, LCMS) available.
  • Soluble in DMSO and water; formulation protocols give clear solutions ≥ 5 mg/mL.
  • Recommended storage: 4°C sealed, away from moisture and light; in solvent store at -80°C for long-term.
  • For research use only; not for human use or sale to patients.

Catalog No. 50-002-85550 Supplier Medchemexpress LLC Supplier No. HY15991A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tenapanor hydrochloride is a potent, orally active sodium/hydrogen exchanger 3 (NHE3) inhibitor supplied for research use. It is provided as a white to off-white solid and is available in solid and solution formats for in vitro and in vivo studies.

  • Potent NHE3 inhibitor for pharmacology and preclinical research.
  • Provided as a white to off-white solid and as solution formulations.
  • Reported purity 99.6% with analytical data (COA, HPLC, LCMS) available.
  • Soluble in DMSO and water; formulation protocols give clear solutions ≥ 5 mg/mL.
  • Recommended storage: 4°C sealed, away from moisture and light; in solvent store at -80°C for long-term.
  • For research use only; not for human use or sale to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.