Learn More
Enzo Life Sciences Tenascin (human) monoclonal antibody (DB7) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLTG60440100
Encompass_Preferred
Clone: DB7. Isotype: IgG1. Immunogen: Purified human tenascin-C. Species reactivity: Human. Applications: IHC, IP, WB. Purity: Partially purified. Formulation: Liquid. In PBS containing 0.1% sodium azide.
Catalog No. 50-201-2222