Learn More
Cayman Chemical Tenatoprazole, Purity: ≥98%, CAS: 113712-98-4, 10 mg
SDP
Catalog No. 502967339
Encompass_Preferred
A proton pump inhibitor, blocking hog gastric H+/K+-ATPase activity with an IC50 value of 6.2 µM; blocks induced gastric acid secretion and ulcer formation in rats and humans
Catalog No. 50-296-7339 Supplier Cayman Chemical Supplier No. 1871710 MG
A proton pump inhibitor, blocking hog gastric H+/K+-ATPase activity with an IC50 value of 6.2 µM; blocks induced gastric acid secretion and ulcer formation in rats and humans