Learn More
Medchemexpress LLC Tenatoprazole | 113712-98-4 | 99.8% | 346.40 | 200 MG
SDP
Catalog No. 5000385651
Encompass_Preferred
Tenatoprazole (TU-199) is an orally active imidazopyridine-based proton pump inhibitor characterized by a prolonged plasma half-life. It inhibits hog gastric H+/K+-ATPase activity and blocks the interaction of ubiquitin with the ESCRT-1 factor Tsg101.
  • Orally active imidazopyridine-based proton pump inhibitor.
  • Characterized by a prolonged plasma half-life.
  • Inhibits hog gastric H+/K+-ATPase activity.
  • Blocks the interaction of ubiquitin with the ESCRT-1 factor Tsg101.
  • Inhibits the production of several enveloped viruses, including EBV.
Catalog No. 50-003-85651 Supplier Medchemexpress LLC Supplier No. HY17421200MG
Tenatoprazole (TU-199) is an orally active imidazopyridine-based proton pump inhibitor characterized by a prolonged plasma half-life. It inhibits hog gastric H+/K+-ATPase activity and blocks the interaction of ubiquitin with the ESCRT-1 factor Tsg101.
  • Orally active imidazopyridine-based proton pump inhibitor.
  • Characterized by a prolonged plasma half-life.
  • Inhibits hog gastric H+/K+-ATPase activity.
  • Blocks the interaction of ubiquitin with the ESCRT-1 factor Tsg101.
  • Inhibits the production of several enveloped viruses, including EBV.