Learn More
Cayman Chemical Tenatoprazole, Purity: ≥98%, CAS: 113712-98-4, 50 mg
SDP
Catalog No. 502965059
Encompass_Preferred
A proton pump inhibitor, blocking hog gastric H+/K+-ATPase activity with an IC50 value of 6.2 µM; blocks induced gastric acid secretion and ulcer formation in rats and humans
Catalog No. 50-296-5059 Supplier Cayman Chemical Supplier No. 1871750 MG
A proton pump inhibitor, blocking hog gastric H+/K+-ATPase activity with an IC50 value of 6.2 µM; blocks induced gastric acid secretion and ulcer formation in rats and humans