Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenax™ Cyphenothrin, MilliporeSigma™ Supelco™

Catalog No. 111019217 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9217 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9217 Supplier MilliporeSigma Supelco Supplier No. 46037100MG
Add to Cart
Add to Cart

Cyphenothrin is classified under the synthetic pyrethroid category of pesticides.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line Tenax
CAS 39515-40-7
Physical Form Neat
Linear Formula C24H25 NO3
MDL Number MFCD01960864
Type Analytical Standard
Quantity 100 mg
Formula Weight 375.46
Grade Analytical Standard

Tenax is a registered trademark of Buchem B.V.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.