Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Tenax™ Porous Polymer Adsorbent, Matrix Tenax TA, 60-80 mesh

Catalog No. 111020776 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0776 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0776 Supplier MilliporeSigma™ Supelco™ Supplier No. 11982
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tenax™ porous polymers, based on 2,6-diphenyl-p-phenylene oxide, is widely used as an adsorbent in both air collection and purge trap applications.

Tenax porous polymers unique structure provides alternate and desirable adsorption/desorption characteristics compared to other porous polymers.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 60/80 Mesh
Packaging Bottle
Quantity 10 g
Surface Area (Metric) ˜35 m2/g
Product Line Tenax
Density ˜0.25 g/mL (free fall density)

Tenax is a registered trademark of Buchem B.V.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.