Learn More
Perkin Elmer US LLC Tenax TA 60/80 - Mesh, 15 g
Catalog No. 502113511
Encompass_Preferred
The approximate analyte volatility range for Tenax TA is n-C7 to n-C26 and the typical analytes are Alkyl benzenes, PAH, PCBs, which is the same as the Tenax GR.Compatible for pumped monitoring and suitable for direct sampling of solids and liquids. Included in the Thermal Desorber Starter Kit (M0413541).
Catalog No. 50-211-3511 Supplier Perkin Elmer US LLC Supplier No. 04978064
The approximate analyte volatility range for Tenax TA is n-C7 to n-C26 and the typical analytes are Alkyl benzenes, PAH, PCBs, which is the same as the Tenax GR.Compatible for pumped monitoring and suitable for direct sampling of solids and liquids. Included in the Thermal Desorber Starter Kit (M0413541).