Learn More
Bellco Glass, Inc. Tenbroeck Homogenizer, 15 mL Glass
SDP
Catalog No. 501945192
Encompass_Preferred
Hollow glass pestle allows addition of ice water for chilling samples during homogenization. All glass construction primarily for manual operation. Interior of the pestle is hollow allowing to reduce heat generated in the grinding process. It can be used with a laboratory stirrer at low speed if a rubber stopper is inserted in the top of the pestle. A short steel rod (not supplied) completes the connection to the collet or Jacobs chuck. CLEARANCE BETWEEN PESTLES & TUBES IS: .004"-.006". CAPACITY: 15ml. STOPPER SIZE: #1.
Catalog No. 50-194-5192 Supplier Bellco Glass, Inc. Supplier No. 198210015
Hollow glass pestle allows addition of ice water for chilling samples during homogenization. All glass construction primarily for manual operation. Interior of the pestle is hollow allowing to reduce heat generated in the grinding process. It can be used with a laboratory stirrer at low speed if a rubber stopper is inserted in the top of the pestle. A short steel rod (not supplied) completes the connection to the collet or Jacobs chuck. CLEARANCE BETWEEN PESTLES & TUBES IS: .004"-.006". CAPACITY: 15ml. STOPPER SIZE: #1.