Learn More
Bellco Glass, Inc. Tenbroeck Homogenizer, 2mL Glass
SDP
Catalog No. 501945190
Encompass_Preferred
Hollow glass pestle allows addition of ice water for chilling samples during homogenization. All glass construction primarily for manual operation. Interior of the pestle is hollow allowing to reduce heat generated in the grinding process. It can be used with a laboratory stirrer at low speed if a rubber stopper is inserted in the top of the pestle. A short steel rod (not supplied) completes the connection to the collet or Jacobs chuck. TOOLED PESTLE FOR USE WITH RUBBER STOPPER (NOT SUPPLIED). CLEARANCE BETWEEN PESTLES & TUBES IS; .004" TO .006".
Catalog No. 50-194-5190 Supplier Bellco Glass, Inc. Supplier No. 198210002
Hollow glass pestle allows addition of ice water for chilling samples during homogenization. All glass construction primarily for manual operation. Interior of the pestle is hollow allowing to reduce heat generated in the grinding process. It can be used with a laboratory stirrer at low speed if a rubber stopper is inserted in the top of the pestle. A short steel rod (not supplied) completes the connection to the collet or Jacobs chuck. TOOLED PESTLE FOR USE WITH RUBBER STOPPER (NOT SUPPLIED). CLEARANCE BETWEEN PESTLES & TUBES IS; .004" TO .006".