Learn More
Electron Microscopy Sciences Tenbroeck Tissue Grinder, 40 ml, 32 mm x 140 mm Mortor, 320 mm OAL
SDP
Catalog No. 501900368
Encompass_Preferred
Glass Tenbroeck tissue grinders are ideal for grinding liver, heart, and intestine tissue. The grinders are precision made and have interchangeable pestles and tubes so there is no need to keep the components matched and parts are easily replacable. The clearance between pestle and tube is 0.0035 to 0.0065" (0.09 – 0.16 mm). While this grinder is designed for hand operation, it can be slow speed motor-driven by inserting a rubber stopper (side 00) and metal rod into pestle. This unit also features an expanded reservoir and pouring lip. Made from Wheaton 33 borosilicate glass. The hollow pestle can be pack with ice. Autoclavable.
Size: 40 ml
Mortor: 32 mm x 140 mm
OAL: 320 mm
Catalog No. 50-190-0368 Supplier Electron Microscopy Sciences Supplier No. 6479840
Glass Tenbroeck tissue grinders are ideal for grinding liver, heart, and intestine tissue. The grinders are precision made and have interchangeable pestles and tubes so there is no need to keep the components matched and parts are easily replacable. The clearance between pestle and tube is 0.0035 to 0.0065" (0.09 – 0.16 mm). While this grinder is designed for hand operation, it can be slow speed motor-driven by inserting a rubber stopper (side 00) and metal rod into pestle. This unit also features an expanded reservoir and pouring lip. Made from Wheaton 33 borosilicate glass. The hollow pestle can be pack with ice. Autoclavable.
Size: 40 ml
Mortor: 32 mm x 140 mm
OAL: 320 mm