Learn More
Selleck Chemical LLC Teneligliptin hydrobromide S4636-20mg
SDP
Catalog No. 503108294
Encompass_Preferred
Teneligliptin (Teneligliptin hydrobromide anhydrous) is a novel potent and long-lasting dipeptidyl peptidase-4 inhibitor competitively inhibited human plasma rat plasma and human recombinant DPP-4 in vitro with IC50 values of approximately 1 nM
Catalog No. 50-310-8294 Supplier Selleck Chemical LLC Supplier No. S463620MG
Teneligliptin (Teneligliptin hydrobromide anhydrous) is a novel potent and long-lasting dipeptidyl peptidase-4 inhibitor competitively inhibited human plasma rat plasma and human recombinant DPP-4 in vitro with IC50 values of approximately 1 nM