Learn More
Medchemexpress LLC Teneligliptin hydrobromide | 906093-29-6 | 100.0% | 628.86 | 5 MG
SDP
Catalog No. 5000361287
Encompass_Preferred
Teneligliptin hydrobromide is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor with IC50 values of 0.37 and 0.29 nM for human and rat DPP-4, respectively. It improves blood glucose levels and is used in research related to type 2 diabetes mellitus.
  • Orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor.
  • Improves blood glucose levels.
  • Used in research related to type 2 diabetes mellitus.
  • Prevents activation of NLRP3 inflammasome.
  • Protects against hypoxia/reoxygenation-induced endothelial cell injury.
  • Inhibits lipopolysaccharide-induced cytotoxicity and inflammation.
Catalog No. 50-003-61287 Supplier Medchemexpress LLC Supplier No. HY14806A5MG
Teneligliptin hydrobromide is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor with IC50 values of 0.37 and 0.29 nM for human and rat DPP-4, respectively. It improves blood glucose levels and is used in research related to type 2 diabetes mellitus.
  • Orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor.
  • Improves blood glucose levels.
  • Used in research related to type 2 diabetes mellitus.
  • Prevents activation of NLRP3 inflammasome.
  • Protects against hypoxia/reoxygenation-induced endothelial cell injury.
  • Inhibits lipopolysaccharide-induced cytotoxicity and inflammation.