Learn More
Apexbio Technology LLC Teneligliptin hydrobromide 906093-29-6 10mg
SDP
Catalog No. 5000096678
Encompass_Preferred
Teneligliptin hydrobromide (CAS 906093-29-6) is a potent dipeptidyl peptidase-4 (DPP-4) inhibitor exhibiting competitive inhibition of human and rat plasma DPP-4 with an IC50 of approximately 1 nM DPP-4 also known as CD26 is a cell surface protease involved in immunoregulation signal transduction and apoptosis In vitro Teneligliptin hydrobromide reduces reactive oxygen species and modulates cell cycle and apoptotic gene expression in human endothelial cells exposed to high glucose In vivo it ameliorates hepatic steatosis and metabolic disturbances in murine models partly through activation of hepatic AMPK This compound supports studies of metabolic oxidative and inflammatory pathways
Catalog No. 50-000-96678 Supplier Apexbio Technology LLC Supplier No. A386510
Teneligliptin hydrobromide (CAS 906093-29-6) is a potent dipeptidyl peptidase-4 (DPP-4) inhibitor exhibiting competitive inhibition of human and rat plasma DPP-4 with an IC50 of approximately 1 nM DPP-4 also known as CD26 is a cell surface protease involved in immunoregulation signal transduction and apoptosis In vitro Teneligliptin hydrobromide reduces reactive oxygen species and modulates cell cycle and apoptotic gene expression in human endothelial cells exposed to high glucose In vivo it ameliorates hepatic steatosis and metabolic disturbances in murine models partly through activation of hepatic AMPK This compound supports studies of metabolic oxidative and inflammatory pathways