Learn More
Medchemexpress LLC Teneligliptin hydrobromide | 906093-29-6 | 99.8% | 628.86 | 1 ML
SDP
Catalog No. 5000363067
Encompass_Preferred
Teneligliptin hydrobromide is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor, demonstrating IC50s of 0.37 nM for human DPP-4 and 0.29 nM for rat DPP-4. This compound improves blood glucose levels and is utilized in research pertaining to type 2 diabetes mellitus.
  • Prevents activation of the NLRP3 inflammasome and associated injury.
  • Protects against hypoxia/reoxygenation-induced endothelial cell injury by reducing reactive oxygen species.
  • Inhibits inflammation and apoptosis induced by doxorubicin.
  • Ameliorates diabetic polyneuropathy and enhances pancreatic β-cell volume and insulin secretion.
  • Attenuates myocardial hypertrophy, injury, and inflammatory response in diabetic models.
  • Prevents obesity and obesity-related manifestations by boosting energy expenditure.
Catalog No. 50-003-63067 Supplier Medchemexpress LLC Supplier No. HY14806A10MM/1ML
Teneligliptin hydrobromide is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor, demonstrating IC50s of 0.37 nM for human DPP-4 and 0.29 nM for rat DPP-4. This compound improves blood glucose levels and is utilized in research pertaining to type 2 diabetes mellitus.
  • Prevents activation of the NLRP3 inflammasome and associated injury.
  • Protects against hypoxia/reoxygenation-induced endothelial cell injury by reducing reactive oxygen species.
  • Inhibits inflammation and apoptosis induced by doxorubicin.
  • Ameliorates diabetic polyneuropathy and enhances pancreatic β-cell volume and insulin secretion.
  • Attenuates myocardial hypertrophy, injury, and inflammatory response in diabetic models.
  • Prevents obesity and obesity-related manifestations by boosting energy expenditure.