Learn More
Medchemexpress LLC Teneligliptin hydrobromide | 906093-29-6 | 99.99% | 628.86 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14806A50MG
Encompass_Preferred
Teneligliptin hydrobromide is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor. It is used in research related to type 2 diabetes mellitus to improve blood glucose levels. This product is for research use only.
  • Orally active
  • Selective dipeptidyl peptidase 4 (DPP-4) inhibitor
  • Improves blood glucose levels
  • For research use only
Catalog No. 50-003-62835