Learn More
Medchemexpress LLC Teneligliptin hydrobromide hydrate | 1572583-29-9 | 100.0% | 426.58 | 5 MG
SDP
Catalog No. 5000362837
Encompass_Preferred
Teneligliptin hydrobromide hydrate is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor. It improves blood glucose levels and is used in research related to type 2 diabetes mellitus. This compound is known for its ability to inhibit DPP-4 with high potency in both human and rat models.
  • Selective dipeptidyl peptidase 4 (DPP-4) inhibitor
  • Improves blood glucose levels
  • Used in research for type 2 diabetes mellitus
  • Prevents activation of NLRP3 inflammasome and injury
  • Protects against hypoxia/reoxygenation-induced endothelial cell injury
  • Attenuates myocardial hypertrophy in diabetic cardiomyopathy
  • Prevents obesity and related manifestations
Catalog No. 50-003-62837 Supplier Medchemexpress LLC Supplier No. HY14806B5MG
Teneligliptin hydrobromide hydrate is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor. It improves blood glucose levels and is used in research related to type 2 diabetes mellitus. This compound is known for its ability to inhibit DPP-4 with high potency in both human and rat models.
  • Selective dipeptidyl peptidase 4 (DPP-4) inhibitor
  • Improves blood glucose levels
  • Used in research for type 2 diabetes mellitus
  • Prevents activation of NLRP3 inflammasome and injury
  • Protects against hypoxia/reoxygenation-induced endothelial cell injury
  • Attenuates myocardial hypertrophy in diabetic cardiomyopathy
  • Prevents obesity and related manifestations