Learn More
Medchemexpress LLC Teneligliptin (hydrobromide hydrate) | 1572583-29-9 | 99.9% | 426.58 | 50 MG
SDP
Catalog No. 5000363070
Encompass_Preferred
Teneligliptin hydrobromide hydrate is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor. It improves blood glucose levels and is used in research related to type 2 diabetes mellitus.
  • Orally active and selective DPP-4 inhibitor
  • Improves blood glucose levels
  • Research tool for type 2 diabetes mellitus
  • Attenuates diabetic cardiomyopathy
  • Prevents obesity in high-fat diet models
Catalog No. 50-003-63070 Supplier Medchemexpress LLC Supplier No. HY14806B50MG
Teneligliptin hydrobromide hydrate is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor. It improves blood glucose levels and is used in research related to type 2 diabetes mellitus.
  • Orally active and selective DPP-4 inhibitor
  • Improves blood glucose levels
  • Research tool for type 2 diabetes mellitus
  • Attenuates diabetic cardiomyopathy
  • Prevents obesity in high-fat diet models