Learn More
Medchemexpress LLC Teneligliptin hydrobromide hydrate | 1572583-29-9 | 99.97% | 426.58 | 25 MG
SDP
Catalog No. 5000363188
Encompass_Preferred
Teneligliptin hydrobromide hydrate is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor with IC50s of 0.37 and 0.29 nM for human and rat DPP-4, respectively. It improves blood glucose levels and is utilized in research related to type 2 diabetes mellitus.
  • Prevents activation of NLRP3 inflammasome and injury in primary mouse cardiomyocytes
  • Protects against hypoxia/reoxygenation-induced endothelial cell injury
  • Inhibits inflammation and apoptosis in H9c2 cells
  • Attenuates myocardial hypertrophy and injury in diabetic cardiomyopathy mice
  • Prevents obesity and related manifestations in high-fat diet-induced obesity models
Catalog No. 50-003-63188 Supplier Medchemexpress LLC Supplier No. 25MGHY14806B
Add to Cart
Add to Cart
Teneligliptin hydrobromide hydrate is an orally active and selective dipeptidyl peptidase 4 (DPP-4) inhibitor with IC50s of 0.37 and 0.29 nM for human and rat DPP-4, respectively. It improves blood glucose levels and is utilized in research related to type 2 diabetes mellitus.
  • Prevents activation of NLRP3 inflammasome and injury in primary mouse cardiomyocytes
  • Protects against hypoxia/reoxygenation-induced endothelial cell injury
  • Inhibits inflammation and apoptosis in H9c2 cells
  • Attenuates myocardial hypertrophy and injury in diabetic cardiomyopathy mice
  • Prevents obesity and related manifestations in high-fat diet-induced obesity models