Learn More
Medchemexpress LLC Teneligliptin hydrobromide hydrate | 1572583-29-9 | MFCD23098790 | 100.0% | 426.58 g/mol | C22H30N6OS | 10 MG
SDP
Catalog No. 5000287686
Encompass_Preferred
Teneligliptin hydrobromide hydrate is the hydrobromide hydrate salt form of teneligliptin, a selective dipeptidyl peptidase-4 (DPP-4) inhibitor supplied for research use. It is provided as a research-grade small molecule for biochemical and cell-based studies and for preclinical investigations of glucose metabolism and DPP-4 pathways.
  • High chemical purity suitable for research applications.
  • Supplied in small mass packs for precise dosing and stock preparation.
  • Hydrobromide hydrate salt form improves solubility for aqueous assays.
  • Documented molecular weight and formula for experimental planning.
  • Datasheet and batch COA available for quality traceability.
Catalog No. 50-002-87686 Supplier Medchemexpress LLC Supplier No. HY14806B10MG
Teneligliptin hydrobromide hydrate is the hydrobromide hydrate salt form of teneligliptin, a selective dipeptidyl peptidase-4 (DPP-4) inhibitor supplied for research use. It is provided as a research-grade small molecule for biochemical and cell-based studies and for preclinical investigations of glucose metabolism and DPP-4 pathways.
  • High chemical purity suitable for research applications.
  • Supplied in small mass packs for precise dosing and stock preparation.
  • Hydrobromide hydrate salt form improves solubility for aqueous assays.
  • Documented molecular weight and formula for experimental planning.
  • Datasheet and batch COA available for quality traceability.