Learn More
TARGETMOL CHEMICALS INC Teneligliptin hydrobromide
SDP
Catalog No. 502880361
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Teneligliptin hydrobromide (MP-513 (hydrobromide)) is a novel, potent, and long-lasting dipeptidyl peptidase-4 inhibitor; competitively inhibited human plasma, rat plasma, and human recombinant DPP-4 in vitro, with IC50 values of approximately 1 nM. Purity 100%
Catalog No. 50-288-0361 Supplier TARGETMOL CHEMICALS INC Supplier No. T699950MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Teneligliptin hydrobromide (MP-513 (hydrobromide)) is a novel, potent, and long-lasting dipeptidyl peptidase-4 inhibitor; competitively inhibited human plasma, rat plasma, and human recombinant DPP-4 in vitro, with IC50 values of approximately 1 nM. Purity 100%