Learn More
Medchemexpress LLC Teniposide | 29767-20-2 | MFCD00866516 | 98.4% | 656.65 | 200 MG
SDP
Catalog No. 5000348505
Encompass_Preferred
Teniposide is a podophyllotoxin derivative that functions as a topoisomerase II inhibitor and is used as a chemotherapeutic agent.
  • Acts as a topoisomerase II inhibitor
  • Functions as a chemotherapeutic agent
  • Demonstrates antiproliferative activity against various human cancer cell lines, including A549, CWR22R, HeLa, HepG2, MCF7, PC-3, and RPMI 8402
  • Induces apoptosis in Tca8113 cells
  • Causes cell cycle arrest at the G2/M phase in Tca8113 cells
  • Can be formulated into a clear solution for in vivo experiments using various protocols, including 10% DMSO, 40% PEG300, 5% Tween-80, and 45% Saline, or 10% DMSO and 90% Corn Oil
  • Referenced in multiple publications
Catalog No. 50-003-48505 Supplier Medchemexpress LLC Supplier No. HY13761200MG
Teniposide is a podophyllotoxin derivative that functions as a topoisomerase II inhibitor and is used as a chemotherapeutic agent.
  • Acts as a topoisomerase II inhibitor
  • Functions as a chemotherapeutic agent
  • Demonstrates antiproliferative activity against various human cancer cell lines, including A549, CWR22R, HeLa, HepG2, MCF7, PC-3, and RPMI 8402
  • Induces apoptosis in Tca8113 cells
  • Causes cell cycle arrest at the G2/M phase in Tca8113 cells
  • Can be formulated into a clear solution for in vivo experiments using various protocols, including 10% DMSO, 40% PEG300, 5% Tween-80, and 45% Saline, or 10% DMSO and 90% Corn Oil
  • Referenced in multiple publications