Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tenivastatin ammonium salt, ≥97.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013045 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3045 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3045 Supplier MilliporeSigma Supelco Supplier No. 0736325MG
Add to Cart
Add to Cart

Specifications

CAS 139893-43-9
Shipping Condition Wet Ice
Linear Formula C25H40O6 NH3
Synonym (betaR,deltaR,1 S,2 S,6 R,8 S,8 aR)-8-(2,2-Dimethyl-1-oxobutoxy)-1,2,6,7,8,8 A-hexahydro-beta,delta-dihydroxy-2,6-dimethyl-1-naphthaleneheptanoic acid ammonium salt; Simvastatin acid ammonium salt; Simvastatin carboxylic acid ammonium salt; Simvastatin hydroxyl acid ammonium salt
Type Analytical Standard
Quantity 25 mg
Formula Weight 453.61
Percent Purity ≥97.5% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.