Learn More
Cayman Chemical Teneligliptin (hydrobromide hydrate), Purity: ≥98%, CAS: 1572583-29-9, 50 mg
SDP
Catalog No. 502989118
Encompass_Preferred
A DPP-4 inhibitor (IC50s = 0.37 and 0.29 nM for the human and rat enzymes, respectively); selective for DPP-4 over DPP-8 and DPP-9 (IC50s = 260 and 540 nM, respectively, for the human enzymes); inhibits increases in plasma glucose levels in an oral glucose tolerance test in Zucker fatty rats at 0.03-1 mg/kg; reduces body weight, inhibits hepatic steatosis, and decreases plasma insulin levels in a mouse model of high-fat diet-induced obesity at 30 and 60 mg/kg; scavenges hydroxyl radicals in a cell-free assay (IC50 = 0.315 mM); decreases urinary levels of 8-OHdG in a DPP-4-deficient rat model of STZ-induced diabetes at 10 mg/kg per day
Catalog No. 50-298-9118 Supplier Cayman Chemical Supplier No. 2951550 MG
A DPP-4 inhibitor (IC50s = 0.37 and 0.29 nM for the human and rat enzymes, respectively); selective for DPP-4 over DPP-8 and DPP-9 (IC50s = 260 and 540 nM, respectively, for the human enzymes); inhibits increases in plasma glucose levels in an oral glucose tolerance test in Zucker fatty rats at 0.03-1 mg/kg; reduces body weight, inhibits hepatic steatosis, and decreases plasma insulin levels in a mouse model of high-fat diet-induced obesity at 30 and 60 mg/kg; scavenges hydroxyl radicals in a cell-free assay (IC50 = 0.315 mM); decreases urinary levels of 8-OHdG in a DPP-4-deficient rat model of STZ-induced diabetes at 10 mg/kg per day