Learn More
Enterprise Technology Solutions Tennsco Corp. Storage Cabinet,w/4 Adj Shelves,Standard,30"x15"x72",Putty
SDP
Catalog No. 5000791385
Encompass_Preferred
Standard-size, slim storage cabinet features premium-quality steel throughout for maximum rigidity. Dual swing-out, reinforced doors ride on heavy-duty leaf-hinges. Three-point locking system ensures secure storage for bulky materials. Simply turn the polished chrome handle and doors lock at three points for maximum security. Nylon guides help make operation quiet. Cabinet base offers a "box" construction technique to prevent racking. Punched lances in back facilitate ventilation. Raised base keeps contents out of harm's way from cleaning liquids. Storage cabinet also includes four adjustable steel shelves that you can move in 2" increments without tools. Each shelf holds up to 150 lb.
Catalog No. 50-007-91385 Supplier Enterprise Technology Solutions Supplier No. TNN7215CPY
Standard-size, slim storage cabinet features premium-quality steel throughout for maximum rigidity. Dual swing-out, reinforced doors ride on heavy-duty leaf-hinges. Three-point locking system ensures secure storage for bulky materials. Simply turn the polished chrome handle and doors lock at three points for maximum security. Nylon guides help make operation quiet. Cabinet base offers a "box" construction technique to prevent racking. Punched lances in back facilitate ventilation. Raised base keeps contents out of harm's way from cleaning liquids. Storage cabinet also includes four adjustable steel shelves that you can move in 2" increments without tools. Each shelf holds up to 150 lb.