Learn More
Enterprise Technology Solutions Tennsco Corp. Storage Cabinet,w/4AdjShelves,Standard,36"x18"x72",Black
SDP
Catalog No. 5000791135
Encompass_Preferred
Commercial grade steel storage cabinet. Built with premium quality steel and reinforced doors for long-term performance at an attractive price. Polished chrome handle uses three-point keyed locking to lock both doors. Storage cabinet is electrostatically painted with a tough, long lasting powder finish to ensure years of lasting beauty.
Catalog No. 50-007-91135 Supplier Enterprise Technology Solutions Supplier No. TNN7218BLK
Commercial grade steel storage cabinet. Built with premium quality steel and reinforced doors for long-term performance at an attractive price. Polished chrome handle uses three-point keyed locking to lock both doors. Storage cabinet is electrostatically painted with a tough, long lasting powder finish to ensure years of lasting beauty.