Learn More
Selleck Chemical LLC Tenofovir Alafenamide
SDP
Supplier:  Selleck Chemical LLC S7856100MG
Encompass_Preferred
Tenofovir Alafenamide is a prodrug of tenofovir which is a reverse transcriptase inhibitor used to treat HIV and Hepatitis B
Catalog No. 50-136-6397