Learn More
Medchemexpress LLC Tenofovir alafenamide | 379270-37-8 | MFCD23843796 | C21H29N6O5P | 1 ML
SDP
Catalog No. 5000372602
Encompass_Preferred
Tenofovir alafenamide (GS-7340) is an investigational oral proagent of Tenofovir, functioning as an HIV-1 nucleotide reverse transcriptase inhibitor. It is intended for research use only.
  • Investigational oral proagent of Tenofovir
  • Functions as an HIV-1 nucleotide reverse transcriptase inhibitor
  • Exhibits antiviral activity against HIV-1 and HIV-2 isolates
  • Effective in PBMCs with EC50s ranging from 0.1 to 12 nM for HIV-1 isolates
  • Soluble in DMSO (≥ 31 mg/mL) and H2O (6.67 mg/mL with ultrasonication)
Catalog No. 50-003-72602 Supplier Medchemexpress LLC Supplier No. HY1523210MM/1ML
Tenofovir alafenamide (GS-7340) is an investigational oral proagent of Tenofovir, functioning as an HIV-1 nucleotide reverse transcriptase inhibitor. It is intended for research use only.
  • Investigational oral proagent of Tenofovir
  • Functions as an HIV-1 nucleotide reverse transcriptase inhibitor
  • Exhibits antiviral activity against HIV-1 and HIV-2 isolates
  • Effective in PBMCs with EC50s ranging from 0.1 to 12 nM for HIV-1 isolates
  • Soluble in DMSO (≥ 31 mg/mL) and H2O (6.67 mg/mL with ultrasonication)