Learn More
Selleck Chemical LLC Tenofovir alafenamide fumarate S4428-25mg
SDP
Catalog No. 503109324
Encompass_Preferred
Tenofovir alafenamide (TAF GS-7340) fumarate is a prodrug of tenofovir but results in significantly higher intracellular tenofovir concentrations and lower serum levels Tenofovir alafenamide is a novel nucleotide reverse transcriptase inhibitor for the treatment of HIV-1 infection
Catalog No. 50-310-9324 Supplier Selleck Chemical LLC Supplier No. S442825MG
Tenofovir alafenamide (TAF GS-7340) fumarate is a prodrug of tenofovir but results in significantly higher intracellular tenofovir concentrations and lower serum levels Tenofovir alafenamide is a novel nucleotide reverse transcriptase inhibitor for the treatment of HIV-1 infection