Learn More
Medchemexpress LLC Tenofovir amibufenamide | 1571076-26-0 | 99.67% | 490.49 | 100 MG
SDP
Catalog No. 5000350547
Encompass_Preferred
Tenofovir amibufenamide (HS-10234) is a Tenofovir prodrug, functioning as an orally active antiviral agent. It effectively inhibits HBV and is utilized in studies concerning chronic hepatitis B (CHB). This compound also regulates hepatocyte metabolism.
  • Inhibits HBV DNA replication
  • Corrects metabolic disorders caused by HBV infection
  • Regulates hepatocyte metabolism
  • Used for chronic hepatitis B (CHB) study
Catalog No. 50-003-50547 Supplier Medchemexpress LLC Supplier No. HY137453100MG
Tenofovir amibufenamide (HS-10234) is a Tenofovir prodrug, functioning as an orally active antiviral agent. It effectively inhibits HBV and is utilized in studies concerning chronic hepatitis B (CHB). This compound also regulates hepatocyte metabolism.
  • Inhibits HBV DNA replication
  • Corrects metabolic disorders caused by HBV infection
  • Regulates hepatocyte metabolism
  • Used for chronic hepatitis B (CHB) study