Learn More
Medchemexpress LLC Tenofovir amibufenamide | 1571076-26-0 | 99.7% | C22H31N6O5P | 10MG
SDP
Catalog No. 5000260417
Encompass_Preferred
Tenofovir amibufenamide is an orally active tenofovir prodrug used in antiviral research to inhibit hepatitis B virus (HBV). It is applied in preclinical studies of chronic hepatitis B to evaluate tenofovir-based antiviral activity and pharmacology.
  • Orally active tenofovir prodrug suitable for in vivo and in vitro studies.
  • Demonstrated inhibition of hepatitis B virus replication.
  • Used in chronic hepatitis B research applications.
  • High purity (99.7%).
  • Molecular formula C22H31N6O5P, molecular weight 490.49 g/mol.
  • Available in small research-scale quantities for preclinical use.
Catalog No. 50-002-60417 Supplier Medchemexpress LLC Supplier No. HY13745310MG
Tenofovir amibufenamide is an orally active tenofovir prodrug used in antiviral research to inhibit hepatitis B virus (HBV). It is applied in preclinical studies of chronic hepatitis B to evaluate tenofovir-based antiviral activity and pharmacology.
  • Orally active tenofovir prodrug suitable for in vivo and in vitro studies.
  • Demonstrated inhibition of hepatitis B virus replication.
  • Used in chronic hepatitis B research applications.
  • High purity (99.7%).
  • Molecular formula C22H31N6O5P, molecular weight 490.49 g/mol.
  • Available in small research-scale quantities for preclinical use.