Learn More
Medchemexpress LLC Tenofovir-d6 (racemic) | 1020719-94-1 | MFCD07369651 | 99.8% | 293.25 g/mol | C9H8D6N5O4P | 5 MG
SDP
Catalog No. 5000263462
Encompass_Preferred
(Rac)-Tenofovir-d6 is a deuterium-labeled, racemic form of tenofovir used as a stable isotope reference and analytical standard. It is intended for pharmacokinetic, metabolism, and quantitative bioanalysis studies of tenofovir and related nucleotide reverse transcriptase inhibitors.
  • Deuterium-labeled (d6) racemic tenofovir.
  • High purity and isotopic enrichment for accurate quantitation.
  • Suitable as an internal standard for LC-MS/MS assays.
  • Ideal for pharmacokinetic and metabolic tracing studies.
  • Supplied in milligram-scale quantities for analytical use.
  • Store at -20 °C and protect from light.
Catalog No. 50-002-63462 Supplier Medchemexpress LLC Supplier No. HY113904S5MG
(Rac)-Tenofovir-d6 is a deuterium-labeled, racemic form of tenofovir used as a stable isotope reference and analytical standard. It is intended for pharmacokinetic, metabolism, and quantitative bioanalysis studies of tenofovir and related nucleotide reverse transcriptase inhibitors.
  • Deuterium-labeled (d6) racemic tenofovir.
  • High purity and isotopic enrichment for accurate quantitation.
  • Suitable as an internal standard for LC-MS/MS assays.
  • Ideal for pharmacokinetic and metabolic tracing studies.
  • Supplied in milligram-scale quantities for analytical use.
  • Store at -20 °C and protect from light.