Learn More
Medchemexpress LLC Tenofovir diphosphate disodium | 2738719-07-6 | 99.4% | C9H14N5Na2O10P3 | 10MG
SDP
Catalog No. 5000259092
Encompass_Preferred
Tenofovir diphosphate disodium is the disodium salt of the active intracellular metabolite of tenofovir. It functions as an antiretroviral nucleotide analog and inhibitor of HIV-1 reverse transcriptase, and is provided for in vitro research into viral replication and polymerase activity. The compound is supplied as a solid for laboratory use.
  • High purity (99.4%).
  • Disodium salt form improves aqueous compatibility.
  • Inhibits HIV-1 reverse transcriptase for biochemical assays.
  • Available in small research pack sizes for laboratory experiments.
  • Solid, white to off-white appearance for easy handling.
  • Intended for research use only, not for human consumption.
Catalog No. 50-002-59092 Supplier Medchemexpress LLC Supplier No. HY136548B10MG
Tenofovir diphosphate disodium is the disodium salt of the active intracellular metabolite of tenofovir. It functions as an antiretroviral nucleotide analog and inhibitor of HIV-1 reverse transcriptase, and is provided for in vitro research into viral replication and polymerase activity. The compound is supplied as a solid for laboratory use.
  • High purity (99.4%).
  • Disodium salt form improves aqueous compatibility.
  • Inhibits HIV-1 reverse transcriptase for biochemical assays.
  • Available in small research pack sizes for laboratory experiments.
  • Solid, white to off-white appearance for easy handling.
  • Intended for research use only, not for human consumption.