Learn More
Medchemexpress LLC Tenofovir diphosphate disodium | 2738719-07-6 | 99.42% | 491.14 | 1 MG
SDP
Catalog No. 5000348861
Encompass_Preferred
Tenofovir diphosphate disodium is an antiretroviral agent primarily used for AIDS research. It acts as an inhibitor of HIV reverse transcriptase DNA (Ki = 1.55 μM) and RNA (Ki = 0.022 μM). This compound exhibits minimal inhibitory effect on eukaryotic DNA replication in vitro.
  • Antiretroviral agent
  • Inhibits HIV reverse transcriptase DNA and RNA
  • Used for AIDS research
  • Minimal inhibitory effect on eukaryotic DNA replication in vitro
Catalog No. 50-003-48861 Supplier Medchemexpress LLC Supplier No. HY136548B1MG
Tenofovir diphosphate disodium is an antiretroviral agent primarily used for AIDS research. It acts as an inhibitor of HIV reverse transcriptase DNA (Ki = 1.55 μM) and RNA (Ki = 0.022 μM). This compound exhibits minimal inhibitory effect on eukaryotic DNA replication in vitro.
  • Antiretroviral agent
  • Inhibits HIV reverse transcriptase DNA and RNA
  • Used for AIDS research
  • Minimal inhibitory effect on eukaryotic DNA replication in vitro