Learn More
Medchemexpress LLC HY-13782A 10mg Medchemexpress, Tenofovir (Disoproxil) CAS:201341-05-1 Purity:>98%
SDP
Catalog No. 502081510
Encompass_Preferred
Medchemexpress, HY-13782A 10mg Tenofovir (Disoproxil) CAS:201341-05-1 Tenofovir Disoproxil (Bis(POC)-PMPA) is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1510 Supplier Medchemexpress LLC Supplier No. HY13782A10MG
Medchemexpress, HY-13782A 10mg Tenofovir (Disoproxil) CAS:201341-05-1 Tenofovir Disoproxil (Bis(POC)-PMPA) is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.