Learn More
Medchemexpress LLC Tenofovir Disoproxil | 201341-05-1 | 99.5% | 519.44 | 1 ML
SDP
Catalog No. 5000350982
Encompass_Preferred
Tenofovir Disoproxil is a nucleotide reverse transcriptase inhibitor. It is commonly used in research to treat HIV and chronic Hepatitis B.
  • Nucleotide reverse transcriptase inhibitor
  • Used to treat HIV
  • Used to treat chronic Hepatitis B
  • High purity of 99.5%
  • Soluble in DMSO (≥ 38 mg/mL)
  • Available as a 10 mM solution in DMSO
Catalog No. 50-003-50982 Supplier Medchemexpress LLC Supplier No. HY13782A10MM/1ML
Tenofovir Disoproxil is a nucleotide reverse transcriptase inhibitor. It is commonly used in research to treat HIV and chronic Hepatitis B.
  • Nucleotide reverse transcriptase inhibitor
  • Used to treat HIV
  • Used to treat chronic Hepatitis B
  • High purity of 99.5%
  • Soluble in DMSO (≥ 38 mg/mL)
  • Available as a 10 mM solution in DMSO