Learn More
Cayman Chemical Tenofovir Disoproxil (fumarate), Purity: ≥98%, CAS: 202138-50-9, 5 mg
SDP
Catalog No. 502968223
Encompass_Preferred
A prodrug of the HIV reverse transcriptase and hepatitis B virus polymerase inhibitor, tenofovir; >100-fold greater anti-HIV activity than that of poorly bioavailable tenofovir in a T cell line and primary blood lymphocytes
Catalog No. 50-296-8223 Supplier Cayman Chemical Supplier No. 169225 MG
A prodrug of the HIV reverse transcriptase and hepatitis B virus polymerase inhibitor, tenofovir; >100-fold greater anti-HIV activity than that of poorly bioavailable tenofovir in a T cell line and primary blood lymphocytes