Learn More
Medchemexpress LLC Tenofovir Disoproxil fumarate | 202138-50-9 | 99.65% | 635.51 | 200 MG
SDP
Catalog No. 5000350586
Encompass_Preferred
Tenofovir Disoproxil fumarate is a nucleotide reverse transcriptase inhibitor used to treat HIV and chronic Hepatitis B. It is intended for research use only and not for patient administration. The product is available as a solid, appearing as a white to off-white substance.
  • Exhibited an IC50 of 4.7 μM against HIV-1 in HeLa cells.
  • Showed an IC50 of 5.1 μM against Hepatitis B virus in HepG2 cells.
  • Demonstrated dose-dependent activity during vaginal HIV challenge in BLT mice.
  • Indicated safety and effectiveness in an HBV infection model in woodchucks.
Catalog No. 50-003-50586 Supplier Medchemexpress LLC Supplier No. HY13782200MG
Tenofovir Disoproxil fumarate is a nucleotide reverse transcriptase inhibitor used to treat HIV and chronic Hepatitis B. It is intended for research use only and not for patient administration. The product is available as a solid, appearing as a white to off-white substance.
  • Exhibited an IC50 of 4.7 μM against HIV-1 in HeLa cells.
  • Showed an IC50 of 5.1 μM against Hepatitis B virus in HepG2 cells.
  • Demonstrated dose-dependent activity during vaginal HIV challenge in BLT mice.
  • Indicated safety and effectiveness in an HBV infection model in woodchucks.