Learn More
Selleck Chemical LLC Tenofovir Disoproxil Fumarate 50mg 202138-50-9 GS-1278 Disoproxil Fumarate
SDP
Supplier:  Selleck Chemical LLC S140050MG
Encompass_Preferred
Tenofovir (GS-1278) Disoproxil Fumarate belongs to a class of antiretroviral drugs, it inhibits the activity of HIV reverse transcriptase by competing with the natural substrate deoxyadenosine 5¿-triphosphate and, after incorporation into DNA, by DNA chain termination. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-630-4