Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tenofovir Disoproxil fumarate (Standard) | 202138-50-9 | 99.7% | 635.51 | 50 MG
SDP

Catalog No. 5000351213
Encompass_Preferred

Tenofovir Disoproxil fumarate (Standard) is an analytical standard intended for research and analytical applications. This nucleotide reverse transcriptase inhibitor is utilized in the treatment of HIV and chronic Hepatitis B.

  • Purity of 99.65%
  • Appears as a solid
  • Color ranges from white to off-white
  • Molecular weight of 635.51
  • Chemical formula C23H34N5O14P
  • CAS number 202138-50-9
  • Serves as a reference standard for assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS

Catalog No. 50-003-51213 Supplier Medchemexpress LLC Supplier No. HY13782R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tenofovir Disoproxil fumarate (Standard) is an analytical standard intended for research and analytical applications. This nucleotide reverse transcriptase inhibitor is utilized in the treatment of HIV and chronic Hepatitis B.

  • Purity of 99.65%
  • Appears as a solid
  • Color ranges from white to off-white
  • Molecular weight of 635.51
  • Chemical formula C23H34N5O14P
  • CAS number 202138-50-9
  • Serves as a reference standard for assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.