Learn More
Medchemexpress LLC Tenofovir exalidex | 911208-73-6 | 99.8% | 569.72 | 10 MG
SDP
Catalog No. 5000346646
Encompass_Preferred
Tenofovir exalidex (CMX-157) is a lipid conjugate of the acyclic nucleotide analog Tenofovir, active against both wild-type and antiretroviral drug-resistant HIV strains. It also demonstrates antiviral activity against HBV. This compound is orally available and shows no apparent toxicity.
  • Effective against multidrug nucleoside/nucleotide analog-resistant viruses.
  • Active against all major subtypes of HIV-1 and HIV-2.
  • Orally available with no apparent toxicity.
Catalog No. 50-003-46646 Supplier Medchemexpress LLC Supplier No. HY10901410MG
Tenofovir exalidex (CMX-157) is a lipid conjugate of the acyclic nucleotide analog Tenofovir, active against both wild-type and antiretroviral drug-resistant HIV strains. It also demonstrates antiviral activity against HBV. This compound is orally available and shows no apparent toxicity.
  • Effective against multidrug nucleoside/nucleotide analog-resistant viruses.
  • Active against all major subtypes of HIV-1 and HIV-2.
  • Orally available with no apparent toxicity.