Learn More
Medchemexpress LLC Tenofovir exalidex | 911208-73-6 | MFCD23843795 | 99.8% | 569.7 g/mol | C28H52N5O5P | 5 MG
SDP
Catalog No. 5000263416
Encompass_Preferred
Tenofovir exalidex is a lipid-conjugated prodrug of the nucleotide analog tenofovir supplied as a research and analytical standard. It exhibits potent antiviral activity against wild-type and drug-resistant HIV strains and shows activity against hepatitis B virus, and is provided in small milligram quantities for in vitro and analytical applications.
  • Lipid-conjugated prodrug designed for improved cell delivery and oral availability.
  • Potent activity against wild-type and antiretroviral-resistant HIV strains.
  • Demonstrates antiviral activity against hepatitis B virus (HBV).
  • Provided as a high-purity analytical standard suitable for in vitro assays.
  • Available in small milligram package sizes for research and assay development.
Catalog No. 50-002-63416 Supplier Medchemexpress LLC Supplier No. HY1090145MG
Tenofovir exalidex is a lipid-conjugated prodrug of the nucleotide analog tenofovir supplied as a research and analytical standard. It exhibits potent antiviral activity against wild-type and drug-resistant HIV strains and shows activity against hepatitis B virus, and is provided in small milligram quantities for in vitro and analytical applications.
  • Lipid-conjugated prodrug designed for improved cell delivery and oral availability.
  • Potent activity against wild-type and antiretroviral-resistant HIV strains.
  • Demonstrates antiviral activity against hepatitis B virus (HBV).
  • Provided as a high-purity analytical standard suitable for in vitro assays.
  • Available in small milligram package sizes for research and assay development.