Learn More
Medchemexpress LLC Tenofovir hydrate | 206184-49-8 | MFCD11519982 | 99.9% | 305.23 g/mol | C9H16N5O5P | 5 MG
SDP
Catalog No. 5000223433
Encompass_Preferred
Tenofovir hydrate is the analytical standard form of tenofovir, a nucleotide reverse transcriptase inhibitor used in HIV and chronic hepatitis B research. Supplied as a high-purity hydrate for analytical and method development applications, it is intended for laboratory research use only and not for human consumption.
  • High reported purity (≈99.88%) for reliable analytical results.
  • Monohydrate form with molecular formula C9H16N5O5P and molecular weight 305.23 g/mol.
  • Supplied in small weighed quantities suitable for standards and validation.
  • Suitable as an analytical standard for HPLC and method development.
  • Accompanied by a certificate of analysis for batch traceability.
Catalog No. 50-002-23433 Supplier Medchemexpress LLC Supplier No. HY13910AR5MG
Tenofovir hydrate is the analytical standard form of tenofovir, a nucleotide reverse transcriptase inhibitor used in HIV and chronic hepatitis B research. Supplied as a high-purity hydrate for analytical and method development applications, it is intended for laboratory research use only and not for human consumption.
  • High reported purity (≈99.88%) for reliable analytical results.
  • Monohydrate form with molecular formula C9H16N5O5P and molecular weight 305.23 g/mol.
  • Supplied in small weighed quantities suitable for standards and validation.
  • Suitable as an analytical standard for HPLC and method development.
  • Accompanied by a certificate of analysis for batch traceability.