Learn More
Electron Microscopy Sciences EMS SuperCut Tenotomy Scissors with Curved Blades, 4-1/2"
SDP
Catalog No. 501931415
Encompass_Preferred
Tenotomy scissors have narrow sharp tips with a razor sharp cutting edge. They are helpful with delicate dissection. Blades have a curve to attain the hard to reach delicate tissue or angle dissections. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings
Blade: Curved
OAL: 4-1/2"
This item is not returnable. View return policy
Catalog No. 50-193-1415 Supplier Electron Microscopy Sciences Supplier No. 7299608
Tenotomy scissors have narrow sharp tips with a razor sharp cutting edge. They are helpful with delicate dissection. Blades have a curve to attain the hard to reach delicate tissue or angle dissections. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings
Blade: Curved
OAL: 4-1/2"