Learn More
Med Vet International Tenotome Knife, Sharp, 5cm x 0.5cm, Curved, J0088SC, Each
Catalog No. 5000584985
Encompass_Preferred
Tenotome Knife. Sharp and curved 5cm x 1/2cm blade.
Catalog No. 50-005-84985 Supplier Med Vet International Supplier No. JOR88SC
Tenotome Knife. Sharp and curved 5cm x 1/2cm blade.